Gene Transcripts

Gene Symbol
EML1
Entrez Gene ID
2009
Gene Name
echinoderm microtubule associated protein like 1
Gene Aliases
ELP79, EMAP, EMAPL, HuEMAP
Chromosome Location
Chr. 14: 99737699 - 99942060 on Build GRCh38
UniGene ID
Hs.12451
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008707.1 NP_001008707.1 13, 14 1473
NM_004434.2 NP_004425.2 12, 13 1416
XM_005267397.1 XP_005267454.1 13, 14 1543
XM_005267398.3 XP_005267455.1 13, 14 1476
XM_005267399.4 XP_005267456.2 13, 14 1515
XM_005267400.4 XP_005267457.1 12, 13 1442
XM_011536540.2 XP_011534842.1 12, 13 1489
XM_011536541.1 XP_011534843.1 13, 14 1556
XM_011536542.2 XP_011534844.1 13, 14 1515
XM_017021074.1 XP_016876563.1 12, 13 1486
GenBank mRNA AK023861.1 - 3, 4 1105
AK126980.1 - 14, 15 1429
AK299812.1 - 13, 14 1752
BC032541.1 - 12, 13 1392 5533599
BC033043.1 - 12, 13 1416 4821202
BX247979.1 - 13, 14 1485
U97018.1 - 12, 13 1415

View Full Product Details