Gene Transcripts
- Gene Symbol
- FLOT2
- Entrez Gene ID
- 2319
- Gene Name
- flotillin 2
- Gene Aliases
- ECS-1, ECS1, ESA, ESA1, M17S1
- Chromosome Location
- Chr. 17: 28879339 - 28897728 on Build GRCh38
- UniGene ID
- Hs.514038
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004475.2 |
NP_004466.2
|
3, 4 |
353 |
|
XM_005257950.3 |
XP_005258007.1
|
3, 4 |
384 |
|
XM_017024394.1 |
XP_016879883.1
|
3, 4 |
281 |
|
XM_017024395.1 |
XP_016879884.1
|
4, 5 |
450 |
|
XM_017024396.1 |
XP_016879885.1
|
3, 4 |
287 |
|
GenBank mRNA |
BC003683.1 |
- |
3, 4 |
265 |
|
BC017292.1 |
- |
2, 3 |
175 |
|
BT019477.1 |
- |
1, 2 |
83 |
|
BT019478.1 |
- |
1, 2 |
83 |
|
CA390100.1 |
- |
3, 4 |
353 |
|
CR541784.1 |
- |
1, 2 |
83 |
|
CR542265.1 |
- |
1, 2 |
83 |
|
M60922.1 |
- |
3, 4 |
209 |
|
View Full Product Details