Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142276.1 | NP_001135748.1 | 17 | 3706 | |
NM_001142277.1 | NP_001135749.1 | 16 | 3538 | ||
NM_001142278.1 | NP_001135750.1 | 13 | 3019 | ||
NM_001243299.1 | NP_001230228.1 | 17 | 3624 | ||
NM_001642.2 | NP_001633.1 | 18 | 3742 | ||
NR_024515.1 | 14 | 3204 | |||
NR_024516.1 | 15 | 3276 | |||
GenBank mRNA | AF168956.1 | - | 18 | 3594 | |
AK056883.1 | - | 14 | 3131 | ||
AK122834.1 | - | 3 | 3976 | ||
AK128162.1 | - | 17 | 3617 | ||
BC000373.2 | - | 17 | 3606 | 2820109 | |
BX647107.1 | - | 3 | 5248 | ||
L09209.1 | - | 18 | 3662 | ||
L19597.1 | - | 16 | 3448 | ||
S60099.1 | - | 18 | 3662 | ||
Z22572.1 | - | 18 | 3651 |