Gene Transcripts

Gene Symbol
ALK
Entrez Gene ID
238
Gene Name
anaplastic lymphoma receptor tyrosine kinase
Gene Aliases
CD246, NBLST3
Chromosome Location
Chr. 2: 29192774 - 29921611 on Build GRCh38
UniGene ID
Hs.654469
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004304.4 NP_004295.2 4 2039
GenBank mRNA AB209477.4 - 4 1576
EU788003.1 - 4 1994
EU788004.1 - 4 1994
EU788005.1 - 4 1994
EU788006.1 - 4 1994
EU788007.1 - 4 1994
EU788008.1 - 4 1994
EU788009.1 - 4 1994
EU788010.1 - 4 1994
FJ838786.1 - 4 1994
FJ838787.1 - 4 1994
GU128152.1 - 4 1087
GU128153.1 - 4 1087
GU128154.1 - 4 1087
GU128155.1 - 4 1087
GU128156.1 - 4 1087
GU128157.1 - 4 1087
U62540.1 - 4 1998
U66559.1 - 4 1235

View Full Product Details