Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_014205.3 | NP_055020.1 | 1 | 1307 | |
GenBank mRNA | AF119497.1 | - | 1 | 1267 | |
AK027081.1 | - | 1 | 1242 | ||
AK223333.1 | - | 1 | 1222 | ||
AK223358.1 | - | 1 | 1277 | ||
BC052240.1 | - | 1 | 1270 | 6107352 |