Gene Transcripts

Gene Symbol
PSKH1
Entrez Gene ID
5681
Gene Name
protein serine kinase H1
Gene Aliases
Chromosome Location
Chr. 16: 67893272 - 67929678 on Build GRCh38
UniGene ID
Hs.513683
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006742.2 NP_006733.1 2 922
GenBank mRNA AJ272212.1 - 2 882
AK309272.1 - 2 922
BC062616.1 - 2 893 5188431
M14504.1 - 1 158

View Full Product Details