Gene Transcripts
- Gene Symbol
- MTG1
- Entrez Gene ID
- 92170
- Gene Name
- mitochondrial ribosome associated GTPase 1
- Gene Aliases
- GTP, GTPBP7
- Chromosome Location
- Chr. 10: 133394117 - 133420670 on Build GRCh38
- UniGene ID
- Hs.501578
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138384.2 |
NP_612393.2
|
5, 6 |
535 |
|
GenBank mRNA |
AK074976.1 |
- |
5, 6 |
456 |
|
AK125012.1 |
- |
6, 7 |
822 |
|
AK292515.1 |
- |
5, 6 |
377 |
|
AK308997.1 |
- |
5, 6 |
555 |
|
BC000920.2 |
- |
3, 4 |
244 |
|
BC004409.1 |
- |
5, 6 |
494 |
|
BC026039.1 |
- |
5, 6 |
511 |
5444745 |
BC026725.1 |
- |
5, 6 |
414 |
|
BC035721.1 |
- |
5, 6 |
439 |
5588933 |
View Full Product Details