Gene Transcripts

Gene Symbol
GRK7
Entrez Gene ID
131890
Gene Name
G protein-coupled receptor kinase 7
Gene Aliases
GPRK7
Chromosome Location
Chr. 3: 141763127 - 141817373 on Build GRCh38
UniGene ID
Hs.680654
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139209.2 NP_631948.1 3 1254
XM_017005717.1 XP_016861206.1 5 3996
GenBank mRNA AF282269.1 - 3 1254
AF439409.1 - 3 1307

View Full Product Details