Gene Transcripts
- Gene Symbol
- CERKL
- Entrez Gene ID
- 375298
- Gene Name
- ceramide kinase like
- Gene Aliases
- RP26
- Chromosome Location
- Chr. 2: 181536672 - 181657107 on Build GRCh38
- UniGene ID
- Hs.732358
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001030311.2 |
NP_001025482.1
|
13, 14 |
1714 |
|
NM_001030312.2 |
NP_001025483.1
|
9, 10 |
1297 |
|
NM_001030313.2 |
NP_001025484.1
|
10, 11 |
1429 |
|
NM_001160277.1 |
NP_001153749.1
|
12, 13 |
1582 |
|
NM_201548.4 |
NP_963842.1
|
12, 13 |
1636 |
|
NR_027689.1 |
|
10, 11 |
1440 |
|
NR_027690.1 |
|
11, 12 |
1572 |
|
GenBank mRNA |
AJ640141.1 |
- |
12, 13 |
1553 |
|
AJ697855.1 |
- |
10, 11 |
2341 |
|
AJ697856.1 |
- |
11, 12 |
1471 |
|
AJ697858.1 |
- |
9, 10 |
1329 |
|
AK129976.1 |
- |
9, 10 |
879 |
|
AK293844.1 |
- |
12, 13 |
1583 |
|
AY357073.1 |
- |
12, 13 |
1535 |
|
AY690329.1 |
- |
13, 14 |
1613 |
|
AY690330.1 |
- |
9, 10 |
1196 |
|
AY690331.1 |
- |
10, 11 |
1328 |
|
AY690332.1 |
- |
10, 11 |
1339 |
|
AY690333.1 |
- |
11, 12 |
1471 |
|
BC137498.1 |
- |
12, 13 |
1562 |
9021498 |
BC137499.1 |
- |
12, 13 |
1562 |
9021499 |
HQ426666.1 |
- |
12, 13 |
1442 |
|
HQ426667.1 |
- |
11, 12 |
1378 |
|
HQ426668.1 |
- |
10, 11 |
1246 |
|
HQ426669.1 |
- |
10, 11 |
1235 |
|
HQ426670.1 |
- |
9, 10 |
1103 |
|
HQ426671.1 |
- |
13, 14 |
1442 |
|
HQ426672.1 |
- |
12, 13 |
1364 |
|
HQ426673.1 |
- |
11, 12 |
1300 |
|
HQ426674.1 |
- |
10, 11 |
1168 |
|
HQ426675.1 |
- |
10, 11 |
1157 |
|
HQ426676.1 |
- |
9, 10 |
1025 |
|
HQ426677.1 |
- |
10, 11 |
972 |
|
HQ426678.1 |
- |
13, 14 |
1689 |
|
HQ426679.1 |
- |
10, 11 |
1994 |
|
JF290420.1 |
- |
12, 13 |
1522 |
|
View Full Product Details