Gene Transcripts

Gene Symbol
XPO6
Entrez Gene ID
23214
Gene Name
exportin 6
Gene Aliases
EXP6, RANBP20
Chromosome Location
Chr. 16: 28097976 - 28211918 on Build GRCh38
UniGene ID
Hs.460468
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270940.1 NP_001257869.1 8 1741
NM_015171.3 NP_055986.1 7 1605
XM_005255197.3 XP_005255254.1 5 939
XM_005255198.1 XP_005255255.1 3 1088
XM_005255199.2 XP_005255256.1 1 217
XM_011545772.2 XP_011544074.1 7 1274
GenBank mRNA AB002368.3 - 7 1556
AK026172.1 - 4 749
AK123846.1 - 8 1373
AY026388.1 - 7 1078
BC004403.1 - 1 345
BC014071.2 - 1 345
BC078674.1 - 3 550
BC108286.1 - 3 507
BC130304.1 - 7 1078 40146335
BC144189.1 - 7 1078 9052709

View Full Product Details