Gene Transcripts

Gene Symbol
MOSPD2
Entrez Gene ID
158747
Gene Name
motile sperm domain containing 2
Gene Aliases
Chromosome Location
Chr. X: 14873405 - 14922166 on Build GRCh38
UniGene ID
Hs.744919
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177475.1 NP_001170946.1 4 428
NM_152581.3 NP_689794.1 5 464
XM_005274451.1 XP_005274508.1 5 464
GenBank mRNA AK093075.1 - 4 428
AK315781.1 - 5 405
BC030641.1 - 5 431 4821861
DA708010.1 - 5 464

View Full Product Details