Gene Transcripts

Gene Symbol
CLEC17A
Entrez Gene ID
388512
Gene Name
C-type lectin domain family 17 member A
Gene Aliases
Chromosome Location
Chr. 19: 14580777 - 14612035 on Build GRCh38
UniGene ID
Hs.567933
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204118.1 NP_001191047.1 14 1924
NM_207390.3 NP_997273.3 13 1814
NR_109784.1 14 1987
NR_109785.1 13 1772
XM_017026785.1 XP_016882274.1 15 2468
XM_017026786.1 XP_016882275.1 15 2332
XM_017026787.1 XP_016882276.1 16 2687
XM_017026788.1 XP_016882277.1 15 2444
XM_017026789.1 XP_016882278.1 11 2146
XM_017026790.1 XP_016882279.1 14 2420
XM_017026791.1 XP_016882280.1 13 2269

View Full Product Details