Gene Transcripts

Gene Symbol
NTM-AS1
Entrez Gene ID
101929637
Gene Name
NTM antisense RNA 1
Gene Aliases
C11orf39
Chromosome Location
Chr. 11: 131662129 - 131663556 on Build GRCh38
UniGene ID
Hs.730330
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK127362.1 - 2 3881

View Full Product Details