Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009023.3 | NP_033049.2 | 2 | 463 | |
GenBank mRNA | AK009672.1 | - | 2 | 464 | |
BC111863.1 | - | 2 | 419 | 40046186 | |
BY709325.1 | - | 2 | 464 | ||
J03962.1 | - | 2 | 399 | ||
X15788.1 | - | 2 | 452 |