Gene Transcripts

Gene Symbol
Trim21
Entrez Gene ID
20821
Gene Name
tripartite motif-containing 21
Gene Aliases
Ro52, Ssa1
Chromosome Location
Chr. 7: 102557920 - 102565482 on Build GRCm38
UniGene ID
Mm.321227
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082552.2 NP_001076021.1 2 201
NM_009277.4 NP_033303.3 3 270
GenBank mRNA AK141294.1 - 3 249
AK150484.1 - 2 165
AK152619.1 - 2 165
AK171489.1 - 3 234
BB661457.1 - 3 257
BC010580.1 - 3 210 3584654
CJ051157.1 - 3 272
CT010336.1 - 1 95
L27990.1 - 3 235

View Full Product Details