Gene Transcripts
- Gene Symbol
- Ranbp2
- Entrez Gene ID
- 19386
- Gene Name
- RAN binding protein 2
- Gene Aliases
- A430087B05Rik, AI256741, NUP358
- Chromosome Location
- Chr. 10: 58446852 - 58494155 on Build GRCm38
- UniGene ID
- Mm.431695
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011240.3 |
NP_035370.2
|
12 |
1805 |
|
GenBank mRNA |
AK145306.1 |
- |
12 |
1736 |
|
View Full Product Details