Gene Transcripts

Gene Symbol
Sema3e
Entrez Gene ID
20349
Gene Name
sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3E
Gene Aliases
6430702L12, AA408817, Semah, mKIAA0331
Chromosome Location
Chr. 5: 14025276 - 14256689 on Build GRCm38
UniGene ID
Mm.134093
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011348.2 NP_035478.2 17 4240
GenBank mRNA AK129116.1 - 12 3005
AK141295.1 - 16 4963
Z93947.1 - 17 4220

View Full Product Details