Gene Transcripts

Gene Symbol
Shcbp1
Entrez Gene ID
20419
Gene Name
Shc SH2-domain binding protein 1
Gene Aliases
mPAL
Chromosome Location
Chr. 8: 4735980 - 4779534 on Build GRCm38
UniGene ID
Mm.37801
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011369.2 NP_035499.1 2 158
GenBank mRNA AF017152.1 - 2 158
AK051205.1 - 2 192
AK052944.1 - 2 184
AK137496.1 - 2 174
AK144966.1 - 2 181
AK149580.1 - 2 175
AK162943.1 - 2 172
BC070455.1 - 2 178 30606785

View Full Product Details