Gene Transcripts

Gene Symbol
Rheb
Entrez Gene ID
19744
Gene Name
Ras homolog enriched in brain
Gene Aliases
Rheb1
Chromosome Location
Chr. 5: 24802823 - 24842361 on Build GRCm38
UniGene ID
Mm.319175
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053075.3 NP_444305.2 2, 3 246
GenBank mRNA AB039919.1 - 2, 3 296
AK151213.1 - 2, 3 278
AK158213.1 - 318
AY197373.1 - 2, 3 246
BC012273.1 - 2, 3 263 3710454

View Full Product Details