Gene Transcripts

Gene Symbol
Golga3
Entrez Gene ID
269682
Gene Name
golgi autoantigen, golgin subfamily a, 3
Gene Aliases
5330413L04, 5430416E01Rik, AI449376, AW490576, G1-499-14, Mea-2, Mea2, repro27
Chromosome Location
Chr. 5: 110176659 - 110226470 on Build GRCm38
UniGene ID
Mm.393008
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008146.3 NP_032172.3 5 884
GenBank mRNA AK133623.1 - 5 823
BC053002.1 - 5 884 5709074
D78270.2 - 5 690

View Full Product Details