Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153176.4 | NP_694816.3 | 5 | 746 | |
GenBank mRNA | AF512565.1 | - | 5 | 778 | |
AF547215.1 | - | 5 | 1075 | ||
AK145540.1 | - | 5 | 747 | ||
AK171647.1 | - | 4 | 645 | ||
BC024986.1 | - | 3 | 401 | ||
BC051051.1 | - | 5 | 680 | ||
BC055488.1 | - | 591 | |||
BC138141.1 | - | 5 | 973 | 8861160 |