Gene Transcripts

Gene Symbol
HS6ST1
Entrez Gene ID
9394
Gene Name
heparan sulfate 6-O-sulfotransferase 1
Gene Aliases
HH15, HS6ST
Chromosome Location
Chr. 2: 128265480 - 128318597 on Build GRCh38
UniGene ID
Hs.512841
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004807.2 NP_004798.3 2 1026
GenBank mRNA AB006179.1 - 2 1100
AK293724.1 - 2 483
AK295898.1 - 2 868
BC001196.1 - 1 275
BC096239.1 - 2 1002 40005544
BC096240.1 - 2 1002 40005547
BC099638.3 - 2 1002 40005546
BC099639.1 - 2 1002 40005548

View Full Product Details