Gene Transcripts

Gene Symbol
Catsperb
Entrez Gene ID
271036
Gene Name
catsper channel auxiliary subunit beta
Gene Aliases
4931410B03, 4932415G16Rik
Chromosome Location
Chr. 12: 101404645 - 101626009 on Build GRCm38
UniGene ID
Mm.84218
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173023.2 NP_766611.2 10, 11 990
GenBank mRNA AK029843.1 - 10, 11 991
AK030010.1 - 10, 11 981
BC132479.1 - 10, 11 911 40130756
BC145000.1 - 10, 11 911
EF199807.1 - 9, 10 871

View Full Product Details