Gene Transcripts

Gene Symbol
Klhdc7a
Entrez Gene ID
242721
Gene Name
kelch domain containing 7A
Gene Aliases
B230308G19Rik
Chromosome Location
Chr. 4: 139962173 - 139968026 on Build GRCm38
UniGene ID
Mm.60400
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173427.2 NP_775603.2 1 1249
GenBank mRNA AK045732.1 - 1 1252
AK050048.1 - 1 836
AK052747.1 - 1 819

View Full Product Details