Gene Transcripts

Gene Symbol
Cnpy1
Entrez Gene ID
269637
Gene Name
canopy 1 homolog (zebrafish)
Gene Aliases
1500012D20Rik, 9630008K15Rik, AI853839
Chromosome Location
Chr. 5: 28200826 - 28237873 on Build GRCm38
UniGene ID
Mm.381479
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175651.4 NP_783582.1 3 2142
GenBank mRNA AK035828.1 - 3 2143

View Full Product Details