Gene Transcripts
- Gene Symbol
- Podxl2
- Entrez Gene ID
- 319655
- Gene Name
- podocalyxin-like 2
- Gene Aliases
- D130074J02Rik, PODLX2
- Chromosome Location
- Chr. 6: 88842557 - 88875053 on Build GRCm38
- UniGene ID
- Mm.302613
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC033384.1 |
- |
|
1992 |
|
View Full Product Details