Gene Transcripts
- Gene Symbol
- Hykk
- Entrez Gene ID
- 235386
- Gene Name
- hydroxylysine kinase 1
- Gene Aliases
- Agphd1, C630028N24Rik
- Chromosome Location
- Chr. 9: 54917290 - 54949924 on Build GRCm38
- UniGene ID
- Mm.259334
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK085527.1 |
- |
5 |
2537 |
|
View Full Product Details