Gene Transcripts
- Gene Symbol
- Sybu
- Entrez Gene ID
- 319613
- Gene Name
- syntabulin (syntaxin-interacting)
- Gene Aliases
- Golsyn
- Chromosome Location
- Chr. 15: 44671856 - 44788063 on Build GRCm38
- UniGene ID
- Mm.207715
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001032727.1 |
NP_001027899.1
|
2, 3 |
666 |
|
NM_001285840.1 |
NP_001272769.1
|
3, 4 |
772 |
|
NM_176998.4 |
NP_795972.2
|
4, 5 |
973 |
|
NM_178765.3 |
NP_848880.2
|
4, 5 |
870 |
|
GenBank mRNA |
AB232447.1 |
- |
4, 5 |
973 |
|
AB232448.1 |
- |
2, 3 |
666 |
|
AB232449.1 |
- |
4, 5 |
870 |
|
AK029072.1 |
- |
3, 4 |
469 |
|
AK030482.1 |
- |
4, 5 |
602 |
|
AK030720.1 |
- |
4, 5 |
602 |
|
AK163432.1 |
- |
4, 5 |
635 |
|
AK220268.1 |
- |
3, 4 |
772 |
|
BC065045.1 |
- |
4, 5 |
867 |
5686153 |
View Full Product Details