Gene Transcripts

Gene Symbol
Nxph2
Entrez Gene ID
18232
Gene Name
neurexophilin 2
Gene Aliases
Chromosome Location
Chr. 2: 23320899 - 23401986 on Build GRCm38
UniGene ID
Mm.131616
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008752.2 NP_032778.1 2 921
GenBank mRNA AK038402.1 - 2 922
BC106863.1 - 2 891 40047329
BC106864.2 - 2 891 40047331

View Full Product Details