Gene Transcripts

Gene Symbol
Edn2
Entrez Gene ID
13615
Gene Name
endothelin 2
Gene Aliases
VIC
Chromosome Location
Chr. 4: 120161424 - 120167360 on Build GRCm38
UniGene ID
Mm.284855
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007902.2 NP_031928.2 5 1114
GenBank mRNA BC037042.1 - 5 1343
X59556.1 - 5 1103

View Full Product Details