Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010266.2 | NP_034396.1 | 7 | 793 | |
GenBank mRNA | AF174583.1 | - | 7 | 719 | |
AF369880.1 | - | 7 | 678 | ||
AF369881.1 | - | 7 | 678 | ||
AF369882.1 | - | 7 | 678 | ||
AF369883.1 | - | 7 | 678 | ||
AF369884.1 | - | 7 | 678 | ||
AK041056.1 | - | 7 | 732 | ||
AK079419.1 | - | 7 | 601 | ||
AK079459.1 | - | 7 | 733 | ||
AK139617.1 | - | 560 | |||
AK157014.1 | - | 7 | 755 | ||
AK173138.1 | - | 7 | 722 | ||
BC076583.1 | - | 7 | 706 | 30620457 |