Gene Transcripts

Gene Symbol
Gdap2
Entrez Gene ID
14547
Gene Name
ganglioside-induced differentiation-associated-protein 2
Gene Aliases
C77050, D3Ertd801e
Chromosome Location
Chr. 3: 100162463 - 100206989 on Build GRCm38
UniGene ID
Mm.279228
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010269.3 NP_034399.1 2 400
GenBank mRNA AK004887.1 - 2 346
AK082958.1 - 2 362
AK146835.1 - 2 390
AK161122.1 - 2 337
AK166799.1 - 2 315
AK166859.1 - 2 314
BC025070.1 - 2 309 3710712
Y17851.1 - 2 398

View Full Product Details