Gene Transcripts

Gene Symbol
PPM1J
Entrez Gene ID
333926
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1J
Gene Aliases
PP2CZ, PP2Czeta, PPP2CZ
Chromosome Location
Chr. 1: 112709994 - 112715328 on Build GRCh38
UniGene ID
Hs.655231
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005167.5 NP_005158.5 7 1106
GenBank mRNA AB451327.1 - 6 461
AB451471.1 - 6 461
AK093270.1 - 7 1086
AL133644.1 - 8 1870
BC011803.2 - 7 1030
BC064628.1 - 7 1126
BC073828.1 - 7 1098 5244363

View Full Product Details