Gene Transcripts

Gene Symbol
Icam2
Entrez Gene ID
15896
Gene Name
intercellular adhesion molecule 2
Gene Aliases
CD102, Icam-2, Ly-60
Chromosome Location
Chr. 11: 106377656 - 106388122 on Build GRCm38
UniGene ID
Mm.394
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010494.1 NP_034624.1 3 711
GenBank mRNA AK007801.1 - 3 646
BC039128.1 - 3 643 3962648
BC039970.1 - 3 663 4487248
X65493.1 - 3 711

View Full Product Details