Gene Transcripts

Gene Symbol
Gfpt2
Entrez Gene ID
14584
Gene Name
glutamine fructose-6-phosphate transaminase 2
Gene Aliases
AI480523, GFAT2
Chromosome Location
Chr. 11: 49794155 - 49838620 on Build GRCm38
UniGene ID
Mm.24402
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013529.3 NP_038557.1 10, 11 1048
GenBank mRNA AB016780.1 - 10, 11 1014
AK132821.1 - 9, 10 894
BC031928.1 - 10, 11 1016 4167189

View Full Product Details