Gene Transcripts

Gene Symbol
Hebp1
Entrez Gene ID
15199
Gene Name
heme binding protein 1
Gene Aliases
Hebp
Chromosome Location
Chr. 6: 135137519 - 135168215 on Build GRCm38
UniGene ID
Mm.378937
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013546.2 NP_038574.2 4 693
GenBank mRNA AB013095.1 - 4 632
AF117613.1 - 4 681
AK008919.1 - 4 614
BC012654.1 - 4 617 4216585

View Full Product Details