Gene Transcripts

Gene Symbol
Igtp
Entrez Gene ID
16145
Gene Name
interferon gamma induced GTPase
Gene Aliases
AW558444, Irgm3
Chromosome Location
Chr. 11: 58199556 - 58207592 on Build GRCm38
UniGene ID
Mm.33902
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018738.4 NP_061208.3 3 532
GenBank mRNA AK002843.1 - 3 470
AK088315.1 - 3 531
AK149817.1 - 3 461
AK151779.1 - 3 472
AK151860.1 - 3 453
AK152337.1 - 3 503
AK152483.1 - 3 453
AK153436.1 - 3 472
AK165065.1 - 3 501
AK169307.1 - 3 491
AY225483.1 - 2 97
BC132557.1 - 2 160 40130834
BC138607.1 - 2 160 8861627
BC145346.1 - 2 160
U53219.1 - 3 444

View Full Product Details