Gene Transcripts

Gene Symbol
Calb2
Entrez Gene ID
12308
Gene Name
calbindin 2
Gene Aliases
CR
Chromosome Location
Chr. 8: 110142538 - 110168206 on Build GRCm38
UniGene ID
Mm.2755
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007586.1 NP_031612.1 6, 7 550
GenBank mRNA AK032171.1 - 556
BC017646.1 - 6, 7 550 4527074
X73985.1 - 4, 5 239

View Full Product Details