Gene Transcripts

Gene Symbol
Cnn1
Entrez Gene ID
12797
Gene Name
calponin 1
Gene Aliases
CN, CnnI
Chromosome Location
Chr. 9: 22099221 - 22109221 on Build GRCm38
UniGene ID
Mm.4356
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009922.4 NP_034052.3 7 1935
GenBank mRNA AK035548.1 - 7 1908

View Full Product Details