Gene Transcripts

Gene Symbol
Cyp2b9
Entrez Gene ID
13094
Gene Name
cytochrome P450, family 2, subfamily b, polypeptide 9
Gene Aliases
Cyp2b
Chromosome Location
Chr. 7: 26173339 - 26210661 on Build GRCm38
UniGene ID
Mm.14413
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010000.2 NP_034130.1 9 1480
GenBank mRNA BC120525.1 - 9 1477
BC120527.1 - 9 1478 40129450
M21855.1 - 9 1456

View Full Product Details