Gene Transcripts
- Gene Symbol
- Aatk
- Entrez Gene ID
- 11302
- Gene Name
- apoptosis-associated tyrosine kinase
- Gene Aliases
- AATYK, aatyk1, mKIAA0641
- Chromosome Location
- Chr. 11: 120007311 - 120047182 on Build GRCm38
- UniGene ID
- Mm.6826
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001198785.1 |
NP_001185714.1
|
8, 9 |
1072 |
|
NM_001198787.1 |
NP_001185716.1
|
8, 9 |
844 |
|
NM_007377.4 |
NP_031403.2
|
8, 9 |
1355 |
|
GenBank mRNA |
AB093253.2 |
- |
8, 9 |
1355 |
|
AB288871.1 |
- |
8, 9 |
974 |
|
AF011908.1 |
- |
8, 9 |
811 |
|
AK036705.1 |
- |
8, 9 |
846 |
|
AK147239.1 |
- |
5, 6 |
865 |
|
AK155100.1 |
- |
8, 9 |
846 |
|
AY236858.1 |
- |
8, 9 |
850 |
|
AY236859.1 |
- |
8, 9 |
877 |
|
BC042837.1 |
- |
8, 9 |
1377 |
|
BC052323.1 |
- |
8, 9 |
1377 |
|
BC080846.1 |
- |
8, 9 |
926 |
6856191 |
View Full Product Details