Gene Transcripts

Gene Symbol
Postn
Entrez Gene ID
50706
Gene Name
periostin, osteoblast specific factor
Gene Aliases
A630052E07Rik, AI747096, OSF-2, Osf2, PLF, PN, peri
Chromosome Location
Chr. 3: 54361096 - 54391041 on Build GRCm38
UniGene ID
Mm.236067
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001198765.1 NP_001185694.1 4 347
NM_001198766.1 NP_001185695.1 4 347
NM_015784.3 NP_056599.1 4 347
GenBank mRNA AK030756.1 - 4 346
AK154049.1 - 4 345
AY651928.1 - 4 317
BC007141.1 - 4 340
BC031449.1 - 4 316 4457222
D13664.1 - 4 329
E05655.1 - 4 343
KM117171.1 - 4 311
KM117172.1 - 4 311

View Full Product Details