Gene Transcripts

Gene Symbol
Pdxp
Entrez Gene ID
57028
Gene Name
pyridoxal (pyridoxine, vitamin B6) phosphatase
Gene Aliases
1600027H05Rik, AB041662, PLPP
Chromosome Location
Chr. 15: 78913919 - 78919517 on Build GRCm38
UniGene ID
Mm.263169
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020271.3 NP_064667.2 2 1242
GenBank mRNA AB041662.1 - 2 1138
AK043228.1 - 2 1203
AY366300.1 - 2 1204
BC037010.1 - 2 666
BC046955.1 - 2 1193
BC049107.1 - 2 1242
BC058388.1 - 2 788
BC069982.1 - 2 1014

View Full Product Details