Gene Transcripts

Gene Symbol
Kbtbd4
Entrez Gene ID
67136
Gene Name
kelch repeat and BTB (POZ) domain containing 4
Gene Aliases
2510026C23Rik
Chromosome Location
Chr. 2: 90904724 - 90910561 on Build GRCm38
UniGene ID
Mm.330394
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025991.3 NP_080267.1 4 1050
GenBank mRNA AK010998.1 - 4 1051
AK014373.1 - 4 976
BC025103.1 - 4 955 4481431

View Full Product Details