Gene Transcripts

Gene Symbol
Haghl
Entrez Gene ID
68977
Gene Name
hydroxyacylglutathione hydrolase-like
Gene Aliases
1500017E18Rik, 2810014I23Rik, C330022E15Rik
Chromosome Location
Chr. 17: 25779843 - 25785669 on Build GRCm38
UniGene ID
Mm.29230
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271433.1 NP_001258362.1 7 786
NM_001271434.1 NP_001258363.1 7 786
NM_001271435.1 NP_001258364.1 7 786
NM_001271436.1 NP_001258365.1 6 600
NM_001271437.1 NP_001258366.1 6 595
NM_026897.3 NP_081173.1 6 771
GenBank mRNA AK005274.1 - 6 767
AK012748.1 - 7 783
AK021220.1 - 6 602
AK032999.1 - 4 744
AK034798.1 - 7 761
AK047517.1 - 7 741
AK079632.1 - 4 935
AK080454.1 - 1 1722
AK154248.1 - 6 576
BC005586.1 - 7 732
BC030466.1 - 7 771 5253213
BC083322.1 - 7 725 6820885

View Full Product Details