Gene Transcripts

Gene Symbol
Trmu
Entrez Gene ID
72026
Gene Name
tRNA 5-methylaminomethyl-2-thiouridylate methyltransferase
Gene Aliases
1110005N20Rik, 1600025P05Rik, AI314320, Trmt1
Chromosome Location
Chr. 15: 85879309 - 85897394 on Build GRCm38
UniGene ID
Mm.158211
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028063.2 NP_082339.1 4 502
GenBank mRNA AK005541.1 - 4 502
AK043532.1 - 4 509
AK044059.1 - 4 509
AY349617.1 - 4 509
BC051548.1 - 4 503
BC061026.1 - 4 489 30305575

View Full Product Details