Gene Transcripts

Gene Symbol
Lancl2
Entrez Gene ID
71835
Gene Name
LanC (bacterial lantibiotic synthetase component C)-like 2
Gene Aliases
1700003F10Rik
Chromosome Location
Chr. 6: 57702455 - 57739449 on Build GRCm38
UniGene ID
Mm.274904
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133737.2 NP_598498.1 3 1088
GenBank mRNA AB035967.1 - 3 907
AK005632.1 - 3 457
BC016072.1 - 3 942 4482042

View Full Product Details