Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_134091.2 | NP_598852.2 | 12 | 1510 | |
GenBank mRNA | AK142314.1 | - | 12 | 1478 | |
AK170806.1 | - | 12 | 1477 | ||
AK183939.1 | - | 3 | 329 | ||
AY382616.1 | - | 12 | 1464 | ||
BC018197.1 | - | 12 | 1470 | 4190407 | |
BC024122.1 | - | 12 | 1470 | 5097970 | |
BC025561.1 | - | 12 | 1448 | 5322980 |