Gene Transcripts

Gene Symbol
Triml1
Entrez Gene ID
244448
Gene Name
tripartite motif family-like 1
Gene Aliases
4933403D14, BC050188
Chromosome Location
Chr. 8: 43129806 - 43180391 on Build GRCm38
UniGene ID
Mm.64542
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177742.4 NP_808410.1 1 441
GenBank mRNA AK077092.1 - 1 443
BC141202.1 - 1 420 9055814
BC145531.1 - 1 420 9054094

View Full Product Details