Gene Transcripts

Gene Symbol
Marveld1
Entrez Gene ID
277010
Gene Name
MARVEL (membrane-associating) domain containing 1
Gene Aliases
AI504298, Mrvldc1
Chromosome Location
Chr. 19: 42147389 - 42151703 on Build GRCm38
UniGene ID
Mm.206250
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183195.2 NP_899018.1 1 1209
GenBank mRNA AK081212.1 - 1 892
AK132243.1 - 1 1200
BC054384.1 - 1 1172 6475446

View Full Product Details